Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 3 (MMP3) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09591
Encompass_Preferred
Rat Matrix Metalloproteinase 3 (MMP3) CLIA Kit for detecting MMP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P03957. Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4425