Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 7 (MMP7) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09598
Encompass_Preferred
Rat Matrix Metalloproteinase 7 (MMP7) CLIA Kit for detecting MMP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P50280. Gene ID: 25335. Target Synonyms: MPSL1; MAT; PUMP-1; Matrilysin Uterine; Matrilysin; Matrin; Pump-1 Protease; Uterine Metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4426