Learn More
Biomatik Corporation Human Tyrosine Kinase With Immunoglobulin Like And EGF Like Domains Protein 1 (Tie1) ELISA Kit
Supplier:  Biomatik Corporation EKU08006
Encompass_Preferred
Human Tyrosine Kinase With Immunoglobulin Like And EGF Like Domains Protein 1 (Tie1) ELISA Kit for detecting Tie1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35590. Gene ID: 7075. Target Synonyms: JTK14; Tyrosine Kinase With Immunoglobulin And Epidermal Growth Factor Homology Domains 1; Tyrosine-protein kinase receptor Tie-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6454