Learn More
Biomatik Corporation Rat Interleukin 24 (IL24) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09881
Encompass_Preferred
Rat Interleukin 24 (IL24) CLIA Kit for detecting IL24 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1000pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9JI24. Gene ID: 170819. Target Synonyms: C49A; FISP; IL10B; MDA7; Mob-5; ST16; Mda-7; Melanoma Differentiation Association Protein 7; Suppression Of Tumorigenicity 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4437