Learn More
Biomatik Corporation Human Transglutaminase 2, Tissue (TGM2) ELISA Kit (CLIA), 96 tests
Supplier: Biomatik Corporation EKU09907
Human Transglutaminase 2 (TGM2) CLIA Kit for detecting TGM2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.22-160ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P21980. Gene ID: 7052. Target Synonyms: tTG; TG2; C Polypeptide,Protein-Glutamine-Gamma-Glutamyltransferase; Transglutaminase 2, Tissue; Transglutaminase C; Transglutaminase H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.