Learn More
Biomatik Corporation Human High Mobility Group Protein 1 (HMG1) ELISA Kit (CLIA), 96 tests
Catalog No. 501504443
Encompass_Preferred
The Human High Mobility Group Protein 1 (HMG1) ELISA Kit (CLIA) allows for quantitative determination of High Mobility Group Protein 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 13.7-10000 pg/mL. Sensitivity: 5.1 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P09429. Cross Activity: No significant cross-reactivity or interference between High Mobility Group Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4443 Supplier Biomatik Corporation Supplier No. EKU09950
The Human High Mobility Group Protein 1 (HMG1) ELISA Kit (CLIA) allows for quantitative determination of High Mobility Group Protein 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 13.7-10000 pg/mL. Sensitivity: 5.1 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P09429. Cross Activity: No significant cross-reactivity or interference between High Mobility Group Protein 1 and analogues was observed. Restriction: For Research Use Only.