Learn More
Biomatik Corporation Human Fibulin 3 (FBLN3) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09976
Encompass_Preferred
The Human Fibulin 3 (FBLN3) ELISA Kit (CLIA) allows for quantitative determination of Fibulin 3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.14-100 ng/mL. Sensitivity: 0.06 ng/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q12805. Cross Activity: No significant cross-reactivity or interference between Fibulin 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4444