Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 5B (WNT5B) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09990
Encompass_Preferred
Human Wingless Type MMTV Integration Site Family, Member 5B (WNT5B) CLIA Kit for detecting WNT5B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9H1J7. Gene ID: 81029. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4448