Learn More
Biomatik Corporation Bilirubin (Bb) ELISA Kit
Supplier:  Biomatik Corporation EKU08395
Encompass_Preferred
Bilirubin (Bb) ELISA Kit for detecting Bb in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ug/ml. Sensitivity: 0.45ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Hematoidin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6496