Learn More
Biomatik Corporation Mouse C-Peptide (CP) ELISA Kit
Supplier:  Biomatik Corporation EKU08446
Encompass_Preferred
Mouse C-Peptide (CP) ELISA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.94-400pg/ml. Sensitivity: 1.82pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01325. Gene ID: 16333. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6500