Learn More
Biomatik Corporation Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit (CLIA), 96 tests
Catalog No. 501504452
Encompass_Preferred
The Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit (CLIA) allows for quantitative determination of Programmed Cell Death Protein 1 Ligand 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 27.4-20,000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q9NZQ7. Cross Activity: No significant cross-reactivity or interference between Programmed Cell Death Protein 1 Ligand 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4452 Supplier Biomatik Corporation Supplier No. EKU10048
The Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit (CLIA) allows for quantitative determination of Programmed Cell Death Protein 1 Ligand 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 27.4-20,000 pg/mL. Sensitivity: 11.5 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q9NZQ7. Cross Activity: No significant cross-reactivity or interference between Programmed Cell Death Protein 1 Ligand 1 and analogues was observed. Restriction: For Research Use Only.