Learn More
Biomatik Corporation 1, 5-Anhydroglucitol (1, 5-AG) ELISA Kit, 96 tests
Catalog No. 501504453
Encompass_Preferred
The 1, 5-Anhydroglucitol (1, 5-AG) ELISA Kit allows for quantitative determination of 1, 5-Anhydroglucitol in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 1.9-150 ug/mL. Sensitivity: 0.7 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between 1, 5-Anhydroglucitol and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4453 Supplier Biomatik Corporation Supplier No. EKU10053
The 1, 5-Anhydroglucitol (1, 5-AG) ELISA Kit allows for quantitative determination of 1, 5-Anhydroglucitol in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 1.9-150 ug/mL. Sensitivity: 0.7 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between 1, 5-Anhydroglucitol and analogues was observed. Restriction: For Research Use Only.