Learn More
Biomatik Corporation Human Antidiuretic Hormone (ADH) ELISA Kit, 96 tests
Catalog No. 501504454
Encompass_Preferred
The Human Antidiuretic Hormone (ADH) ELISA Kit allows for quantitative determination of Antidiuretic Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Antidiuretic Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4454 Supplier Biomatik Corporation Supplier No. EKU10063
The Human Antidiuretic Hormone (ADH) ELISA Kit allows for quantitative determination of Antidiuretic Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Antidiuretic Hormone and analogues was observed. Restriction: For Research Use Only.