Learn More
Biomatik Corporation Rat Neuromedin U (NMU) ELISA Kit
Supplier:  Biomatik Corporation EKU08574
Encompass_Preferred
Rat Neuromedin U (NMU) ELISA Kit for detecting NMU in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 49.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P12760. Gene ID: 63887. Target Synonyms: NM-U. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6514