Learn More
Biomatik Corporation Rabbit Apolipoprotein B100 (APOB100) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10066
Encompass_Preferred
Rabbit Apolipoprotein B100 (APOB100) ELISA Kit for detecting APOB100 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 78-5,000ng/ml. Sensitivity: 37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1U9R4. Gene ID: 100009101. Target Synonyms: Apo-B100. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4456