Learn More
Biomatik Corporation Human Atrial Natriuretic Peptide (ANP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10067
Encompass_Preferred
The Human Atrial Natriuretic Peptide (ANP) ELISA Kit allows for quantitative determination of Atrial Natriuretic Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.62 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Atrial Natriuretic Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4457