Learn More
Biomatik Corporation Mouse Brain Natriuretic Peptide (BNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10073
Encompass_Preferred
Mouse Brain Natriuretic Peptide (BNP) ELISA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.84pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P40753. Gene ID: 18158. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4458