Learn More
Biomatik Corporation Rat Brain Natriuretic Peptide (BNP) ELISA Kit, 96 tests
Catalog No. 501504459
Encompass_Preferred
Rat Brain Natriuretic Peptide (BNP) ELISA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13205. Gene ID: 25105. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4459 Supplier Biomatik Corporation Supplier No. EKU10076
Rat Brain Natriuretic Peptide (BNP) ELISA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13205. Gene ID: 25105. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.