Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 15 (FGF15) ELISA Kit
Supplier:  Biomatik Corporation EKU08757
Encompass_Preferred
Rat Fibroblast Growth Factor 15 (FGF15) ELISA Kit for detecting FGF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-200pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8VI81. Gene ID: 170582. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6544