Learn More
Biomatik Corporation 2-Arachidonoylglycerol (2-AG) ELISA Kit
Catalog No. 502286546
Encompass_Preferred
2-Arachidonoylglycerol (2-AG) ELISA Kit for detecting 2-AG in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.70-300ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6546 Supplier Biomatik Corporation Supplier No. EKU08760
2-Arachidonoylglycerol (2-AG) ELISA Kit for detecting 2-AG in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 3.70-300ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.