Learn More
Biomatik Corporation Mouse Cathelicidin Antimicrobial Peptide (CAMP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10086
Encompass_Preferred
Mouse Cathelicidin Antimicrobial Peptide (CAMP) ELISA Kit for detecting CAMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 123.5-10000ng/ml. Sensitivity: 52.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P51437. Gene ID: 12796. Target Synonyms: CAP18; FALL39; HSD26; LL37; 18 kDa Cationic Antimicrobial Protein; Antibacterial protein FALL-39; FALL-39 peptide antibiotic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4462