Learn More
Biomatik Corporation Human Immunoglobulin G1 (IgG1) ELISA Kit
Supplier:  Biomatik Corporation EKU08972
Encompass_Preferred
Human Immunoglobulin G1 (IgG1) ELISA Kit for detecting IgG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.83ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01768. Target Synonyms: IGHG1; Ig Gamma-1 Chain C Region; Immunoglobulin Heavy Constant Gamma 1; G1m Marker; Immunoglobulin Gm1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6557