Learn More
Biomatik Corporation Human Chorionic Gonadotropin Beta Polypeptide (CGb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10090
Encompass_Preferred
The Human Chorionic Gonadotropin Beta Polypeptide (CGb) ELISA Kit allows for quantitative determination of Chorionic Gonadotropin Beta Polypeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-50 ng/mL. Sensitivity: 1.38 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P0DN86. Cross Activity: No significant cross-reactivity or interference between Chorionic Gonadotropin Beta Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4464