Learn More
Biomatik Corporation Mouse Aminopeptidase A (ENPEP) ELISA Kit
Supplier:  Biomatik Corporation EKU09028
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P16406. Target Synonyms: CD249; GluAP; gp160; ATA; EAP; AP-A; Aminopeptidase A; Glutamyl Aminopeptidase. Restriction: For Research Use Only.
Catalog No. 50-228-6566