Learn More
Biomatik Corporation Human/Mouse/Rat Signal Transducer And Activator Of Transcription 3 (STAT3) ELISA Kit
Supplier:  Biomatik Corporation EKU09080
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human/Mouse/Rat. Detection Range: 1.56-100ng/mL. Sensitivity: 0.56ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: APRF; Acute-Phase Response Factor. Restriction: For Research Use Only.
Catalog No. 50-228-6574