Learn More
Biomatik Corporation Human Fibroblast Growth Factor 18 (FGF18) ELISA Kit, 96 tests
Catalog No. 501504468
Encompass_Preferred
The Human Fibroblast Growth Factor 18 (FGF18) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O76093. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4468 Supplier Biomatik Corporation Supplier No. EKU10106
The Human Fibroblast Growth Factor 18 (FGF18) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O76093. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 18 and analogues was observed. Restriction: For Research Use Only.