Learn More
Biomatik Corporation Cattle Growth Hormone (GH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10114
Encompass_Preferred
The Cattle Growth Hormone (GH) ELISA Kit allows for quantitative determination of Growth Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 123.5-10000 pg/mL. Sensitivity: 51.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01246. Cross Activity: No significant cross-reactivity or interference between Growth Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4471