Learn More
Biomatik Corporation Rat Growth Hormone (GH) ELISA Kit, 96 tests
Catalog No. 501504472
Encompass_Preferred
Rat Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 43.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01244. Gene ID: 24391. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4472 Supplier Biomatik Corporation Supplier No. EKU10120
Rat Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 43.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01244. Gene ID: 24391. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.