Learn More
Biomatik Corporation Human Guanylate Cyclase Activator 2A (GUCA2A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10121
Encompass_Preferred
The Human Guanylate Cyclase Activator 2A (GUCA2A) ELISA Kit allows for quantitative determination of Guanylate Cyclase Activator 2A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02747. Cross Activity: No significant cross-reactivity or interference between Guanylate Cyclase Activator 2A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4473