Learn More
Biomatik Corporation Human Autophagy Related Protein 5 (ATG5) ELISA Kit
Supplier:  Biomatik Corporation EKU09301
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.29ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9H1Y0. Target Synonyms: ASP; APG5; APG5L; hAPG5; Apoptosis-specific protein; Autophagy protein 5. Restriction: For Research Use Only.
Catalog No. 50-228-6602