Learn More
Biomatik Corporation Human Hepcidin (Hepc) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10126
Encompass_Preferred
The Human Hepcidin (Hepc) ELISA Kit allows for quantitative determination of Hepcidin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 26.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P81172. Cross Activity: No significant cross-reactivity or interference between Hepcidin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4475