Learn More
Biomatik Corporation Rat Immunoglobulin M (IgM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10135
Encompass_Preferred
Rat Immunoglobulin M (IgM) ELISA Kit for detecting IgM in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.99ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: F1LPW0. Target Synonyms: IGHM; Immunoglobulin Heavy Constant Mu; Ig mu chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4479