Learn More
Biomatik Corporation Rat Myostatin (MSTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10160
Encompass_Preferred
Rat Myostatin (MSTN) ELISA Kit for detecting MSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.227ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35312. Gene ID: 29152. Target Synonyms: GDF8; Growth Differentiation Factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4481