Learn More
Biomatik Corporation Mouse Noggin (NOG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10172
Encompass_Preferred
Mouse Noggin (NOG) ELISA Kit for detecting NOG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97466. Gene ID: 18121. Target Synonyms: SYM1; SYNS1; Synostoses (Multiple) Syndrome 1; Symphalangism 1 (Proximal). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4483