Learn More
Biomatik Corporation Human Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10181
Encompass_Preferred
The Human Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit allows for quantitative determination of Oxidized Low Density Lipoprotein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.23-100 ng/mL. Sensitivity: 0.52 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P04114. Cross Activity: No significant cross-reactivity or interference between Oxidized Low Density Lipoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4486