Learn More
Biomatik Corporation Mouse Parathyroid Hormone Related Protein (PTHrP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10188
Encompass_Preferred
Mouse Parathyroid Hormone Related Protein (PTHrP) ELISA Kit for detecting PTHrP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.35pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P22858. Target Synonyms: PTHLH; HHM; PTHR; PLP; Osteostatin; Parathyroid Hormone Like Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4487