Learn More
Biomatik Corporation Human Procollagen I N-Terminal Propeptide (PINP) ELISA Kit, 96 tests
Catalog No. 501504488
Encompass_Preferred
The Human Procollagen I N-Terminal Propeptide (PINP) ELISA Kit allows for quantitative determination of Procollagen I N-Terminal Propeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.91 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P02452. Cross Activity: No significant cross-reactivity or interference between Procollagen I N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4488 Supplier Biomatik Corporation Supplier No. EKU10200
The Human Procollagen I N-Terminal Propeptide (PINP) ELISA Kit allows for quantitative determination of Procollagen I N-Terminal Propeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.91 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P02452. Cross Activity: No significant cross-reactivity or interference between Procollagen I N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.