Learn More
Biomatik Corporation Rat Prolactin (PRL) ELISA Kit, 96 tests
Catalog No. 501504493
Encompass_Preferred
Rat Prolactin (PRL) ELISA Kit for detecting PRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01237. Gene ID: 24683. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4493 Supplier Biomatik Corporation Supplier No. EKU10210
Rat Prolactin (PRL) ELISA Kit for detecting PRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01237. Gene ID: 24683. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.