Learn More
Biomatik Corporation Human Rho Associated Coiled Coil Containing Protein Kinase 2 (Rock2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10215
Encompass_Preferred
Human Rho Associated Coiled Coil Containing Protein Kinase 2 (Rock2) ELISA Kit for detecting Rock2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75116. Gene ID: 9475. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4494