Learn More
Biomatik Corporation Mouse Somatostatin (SST) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10227
Encompass_Preferred
Mouse Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60041. Gene ID: 20604. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4497