Learn More
Biomatik Corporation Human Tissue Factor Pathway Inhibitor (TFPI) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10321
Encompass_Preferred
Human Tissue Factor Pathway Inhibitor (TFPI) High-sensitive ELISA Kit for detecting TFPI in plasma, urine, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10646. Gene ID: 7035. Target Synonyms: TFI; EPI; LACI; TFPI1; Lipoprotein-Associated Coagulation Inhibitor; Extrinsic pathway inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6695