Learn More
Biomatik Corporation Rat Ubiquitin Carboxyl Terminal Hydrolase L1 (UCHL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10252
Encompass_Preferred
Rat Ubiquitin Carboxyl Terminal Hydrolase L1 (UCHL1) ELISA Kit for detecting UCHL1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00981. Gene ID: 29545. Target Synonyms: PARK5; PGP9.5; Uch-L1; Neuron cytoplasmic protein 9.5; Ubiquitin thioesterase L1; Ubiquitin carboxyl-terminal hydrolase isozyme L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4501