Learn More
Biomatik Corporation Human Interferon Gamma (IFNg) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10640
Encompass_Preferred
Human Interferon Gamma (IFNg) Anti-antigen ELISA Kit for detecting IFNg in serum. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P01579. Gene ID: 3458. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6726