Learn More
Biomatik Corporation Chondroitin Sulfate (CS) ELISA Kit
Supplier:  Biomatik Corporation EKU10718
Encompass_Preferred
Chondroitin Sulfate (CS) ELISA Kit for detecting CS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 39.06-10,000pg/ml. Sensitivity: 16.22pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6749