Learn More
Biomatik Corporation High Sensitive Mouse Matrix Metalloproteinase 13 (MMP13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10300
Encompass_Preferred
The High Sensitive Mouse Matrix Metalloproteinase 13 (MMP13) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 13 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P33435. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 13 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4511