Learn More
Biomatik Corporation High Sensitive Mouse Microtubule Associated Protein Tau (MAPt) ELISA Kit, 96 tests
Catalog No. 501504513
Encompass_Preferred
The High Sensitive Mouse Microtubule Associated Protein Tau (MAPt) ELISA Kit allows for quantitative determination of Microtubule Associated Protein Tau in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10637. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein Tau and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4513 Supplier Biomatik Corporation Supplier No. EKU10303
The High Sensitive Mouse Microtubule Associated Protein Tau (MAPt) ELISA Kit allows for quantitative determination of Microtubule Associated Protein Tau in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10637. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein Tau and analogues was observed. Restriction: For Research Use Only.