Learn More
Biomatik Corporation Rabbit Cross Linked C-Telopeptide Of Type II Collagen (CTXII) ELISA Kit
Supplier:  Biomatik Corporation EKU10756
Encompass_Preferred
Rabbit Cross Linked C-Telopeptide Of Type II Collagen (CTXII) ELISA Kit for detecting CTXII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 98.8-8,000pg/ml. Sensitivity: 36.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: CTX-II; CTX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6758