Learn More
Biomatik Corporation High Sensitive Rat Microtubule Associated Protein Tau (MAPt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10304
Encompass_Preferred
The High Sensitive Rat Microtubule Associated Protein Tau (MAPt) ELISA Kit allows for quantitative determination of Microtubule Associated Protein Tau in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19332. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein Tau and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4514