Learn More
Biomatik Corporation Human Cytochrome C (CYCS) ELISA Kit
Supplier:  Biomatik Corporation EKU10779
Encompass_Preferred
Human Cytochrome C (CYCS) ELISA Kit for detecting CYCS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P99999. Gene ID: 54205. Target Synonyms: Cyt-C; HCS; CYC; CytC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6762