Learn More
Biomatik Corporation High Sensitive Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10307
Encompass_Preferred
The High Sensitive Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit allows for quantitative determination of Neurofilament, Light Polypeptide in Serum, Plasma, cerebrospinal fluid and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.63 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08551. Cross Activity: No significant cross-reactivity or interference between Neurofilament, Light Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4517